VWR® AVS TITRINORM Buffer Solutions, 20 °C
4,00, ±0,02, Phthalate, Colourless, ISO 17034, Bag-in-box, 5 L
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.
Reference temperature 20 °C
- True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 10,00); ±0,05 for pH 11,00 and pH 12,00 buffer
- Traceable to SRM from NIST
- Batch number and packing date for traceability
- Certificate of Analysis available at website
Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 10,00 and 0,05 pH unit for pH 11 and pH 12,0.
: Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.
: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.
: Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).
Documents
Certifikater
Søg efter partinummer100 ml plastic bottle for liquids DIN 28 (ZPACK13948098) 10L Cubitainer UN (ZPACK13260004) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 250 ml Glass bottle for liquids DIN 28 (ZPACK13949250) 500 ml Plastic bottle for liquids DIN 45 (ZPACK13960500)
Packaging Information
More
P20_VWRC32032.260_SPEC_EN.pdf P20_VWRC32039.261_SPEC_EN.pdf P20_VWRC32039.294_SPEC_EN.pdf P20_VWRC32040.260_SPEC_EN.pdf P20_VWRC32044.290_SPEC_EN.pdf
Product specification
More
SDS_EN_CH_VWRC32031 SDS_EN_CH_VWRC32044 SDS_EN_CH_VWRC32045 SDS_EN_CH_VWRC32045.pdf SDS_EN_CH_VWRC32046
SDS
More
VWR® for Calibrations VWR® pH Buffer Standard Solutions.pdf
Product Literature
Specifikationer
Specifikationer
- Pack type:Bag-in-box
- pH buffer type:Phthalate
- Pk:5 L
- Colour:Colourless
- pH accuracy:±0,02
- pH value:4,00
- Accreditation:ISO 17034



