VWR® AVS TITRINORM Buffer Solutions, 20 °C
Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.
- Reference temperature 20 °C
- True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 11,00); ±0,05 for pH 12,00 buffer
- Traceable to SRM from NIST
- Batch number and packing date for traceability
- Certificate of Analysis available at website
Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 11,00 and 0,05 pH unit for pH 12,0.
: Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.
: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.
: Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).
Documents
100 ml plastic bottle for liquids DIN 28 (ZPACK13948098) 10L Cubitainer UN (ZPACK13260004) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 250 ml Glass bottle for liquids DIN 28 (ZPACK13949250) 500 ml Plastic bottle for liquids DIN 45 (ZPACK13960500)
Emballageoplysninger
P20_VWRC32031.297_SPEC_EN.pdf P20_VWRC32032.260_SPEC_EN.pdf P20_VWRC32032.291_SPEC_EN.pdf P20_VWRC32032.291_SPEC_PL.pdf P20_VWRC32033.294_SPEC_EN.pdf
Produktspecifikation
SDS_EN_CH_VWRC32031 SDS_EN_CH_VWRC32044 SDS_EN_CH_VWRC32045 SDS_EN_CH_VWRC32045.pdf SDS_EN_CH_VWRC32046
SDS
VWR® for Calibrations VWR® pH Buffer Standard Solutions.pdf
Produktlitteratur
- Pack type:Bag-in-box
- pH buffer type:Borate
- Pk:5 L
- Colour:Blue
- pH accuracy:±0,02
- pH value:9,00
- Accreditation:ISO 17034
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



