VWR® AVS TITRINORM Buffer Solutions, 20 °C
12,00, ±0,05, Phosphate, Colourless, -, Plastic bottle, 1 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.
Reference temperature 20 °C
- True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 10,00); ±0,05 for pH 11,00 and pH 12,00 buffer
- Traceable to SRM from NIST
- Batch number and packing date for traceability
- Certificate of Analysis available at website
Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 10,00 and 0,05 pH unit for pH 11 and pH 12,0.
: Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.
: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.
: Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).
Documents
Certificates
Search by Lot Number100 ml plastic bottle for liquids DIN 28 (ZPACK13948098) 10L Cubitainer UN (ZPACK13260004) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 250 ml Glass bottle for liquids DIN 28 (ZPACK13949250) 500 ml Plastic bottle for liquids DIN 45 (ZPACK13960500)
Packaging Information
More
P20_VWRC32039.261_SPEC_EN.pdf spec VWR AVS TITRINORM Buffer Solutions VWRC32031.297 spec VWR AVS TITRINORM Buffer Solutions VWRC32032.260 spec VWR AVS TITRINORM Buffer Solutions VWRC32032.291 spec VWR AVS TITRINORM Buffer Solutions VWRC32033.294
Product specification
More
SDS_EN_CH_VWRC32031 SDS_EN_CH_VWRC32044 SDS_EN_CH_VWRC32045 SDS_EN_CH_VWRC32045.pdf SDS_EN_CH_VWRC32046
SDS
More
VWR® for Calibrations VWR® pH Buffer Standard Solutions.pdf
Product Literature
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Phosphate
- Pk:1 L
- Colour:Colourless
- pH accuracy:±0,05
- pH value:12,00
- Accreditation:-



