Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® Drum Keys
VWR® Drum Keys
Drum keys, plastic, for tightening/opening caps for 20 or 25 L (DIN 61) plastic drums
Catalog # 29548.053
Supplier:   VWR Collection
VWR® Drum Keys
Catalog # 29548.053
Supplier:  VWR Collection
VWR® Drum Keys
VWR® Drum Keys
Drum keys, plastic, for tightening/opening caps for 20 or 25 L (DIN 61) plastic drums
Price
63,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
A selection of tools to assist in opening IBCs, 200 litre drums, plus bottles and other containers with DIN 32, 45, 54 and 80 caps.
Specifications
  • Description:
    Drum keys, plastic, for tightening/opening caps for 20 or 25 L (DIN 61) plastic drums