Order Entry
Denmark
Orders LinkContactUsLinkComponent
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×143 mm (Ø×H), un-numbered, 300 ml
Catalog # WHEA227498
Supplier:   DWK Life Sciences
BOD bottles, WHEATON®
Catalog # WHEA227498
Supplier:  DWK Life Sciences
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×143 mm (Ø×H), un-numbered, 300 ml
Price
4.800,00 kr
Log in for availability
1 * 24 Items
Product Details & Documents
Made of Type I borosilicate glass.
  • Available with or without stopper
  • With white labeling area
Specifications
  • Capacity:
    300 ml
  • Description:
    BOD bottles, with bar codes, borosilicate glass, 69×143 mm (Ø×H), un-numbered