Sodium L(+)-ascorbate ≥99%, TECHNICAL
Plastic bottle for solids, 1 kg
Supplier: VWR Chemicals
Sodium L(+)-ascorbate ≥99%, TECHNICAL
Catalog # 27688.292
Supplier: VWR Chemicals
CAS Number: 134-03-2
Jump to:
Product Details & Documents
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS Number:134-03-2
- Formula:C₆H₇NaO₆
- MDL Number:MFCD00082340
- Molecular Weight:198.11 g/mol
- Melting Pt:219 °C
- Density:1.799 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-126-1
Documents
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC27688.235 spec_VWRC27688.292
Product specification
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99 %
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg



