Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Mannitol, GPR RECTAPUR®

D-(-)-Mannitol, GPR RECTAPUR®

Plastic bucket, 5 kg
Catalog # 25313.363
Supplier:   VWR Chemicals
D-(-)-Mannitol, GPR RECTAPUR®
Catalog # 25313.363
Supplier:  VWR Chemicals
CAS Number:  69-65-8
D-(-)-Mannitol, GPR RECTAPUR®
D-(-)-Mannitol, GPR RECTAPUR®

Plastic bucket, 5 kg

CAS Number:  69-65-8
Price
2.640,00 kr
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specification Test Results

  • Melting point:
    165 - 170 °C
  • Specific optical rotation (8 %; borate):
    23 - 24.5 °
  • Ignition residue (SO4):
    Max. 0.2 %
  • Loss on drying (100°C):
    Max. 0.5 %
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 0.05 %

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.