Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Mannitol, GPR RECTAPUR®

D-(-)-Mannitol, GPR RECTAPUR®

Plastic bottle for solids, 1 kg
Catalog # 25313.294
Supplier:   VWR Chemicals
D-(-)-Mannitol, GPR RECTAPUR®
Catalog # 25313.294
Supplier:  VWR Chemicals
CAS Number:  69-65-8
D-(-)-Mannitol, GPR RECTAPUR®
D-(-)-Mannitol, GPR RECTAPUR®

Plastic bottle for solids, 1 kg

CAS Number:  69-65-8
Price
784,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specification Test Results

  • Melting point:
    165 - 170 °C
  • Specific optical rotation (8 %; borate):
    23 - 24.5 °
  • Ignition residue (SO4):
    Max. 0.2 %
  • Loss on drying (100°C):
    Max. 0.5 %
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 0.05 %

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications