Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

n-Hexane ≥95%, TECHNICAL

Metal drum, 25 L
Catalog # 24611.468
Flammable
Health
Toxic
Environment
Supplier:   VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Catalog # 24611.468
Supplier:  VWR Chemicals
CAS Number:  110-54-3
n-Hexane ≥95%, TECHNICAL
n-Hexane ≥95%, TECHNICAL

Metal drum, 25 L

CAS Number:  110-54-3
Price
2.210,00 kr
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94,3 °C
  • Density:0.659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
  • REACH:01-2119480412-44
Specifications

Specification Test Results

  • Assay:
    Min. 95 %
  • Boiling point:
    68 - 70 °C
  • Benzene:
    Max. 100 ppm
  • Evaporation residue:
    Max. 20 ppm

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.