Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories

Accessories

Gelatin filters, wavelength 680 nm, bandpass ± 5 nm
Catalog # 633-0119
Accessories
Catalog # 633-0119
Accessories

Gelatin filters, wavelength 680 nm, bandpass ± 5 nm

Price
3.640,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Gelatin filters, wavelength 680 nm, bandpass ± 5 nm
  • Seq:
    05