Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories

Accessories

Gelatin filters, wavelength 510 nm, bandpass ± 5 nm
Catalog # 633-0116
Accessories
Catalog # 633-0116
Accessories

Gelatin filters, wavelength 510 nm, bandpass ± 5 nm

Price
3.640,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Gelatin filters, wavelength 510 nm, bandpass ± 5 nm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.