0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
Orders Link
ContactUsLinkComponent
Trays/Dishes/Pots
Instrument Trays/Dishes
Instrument trays, rectangular
Instrument trays, rectangular
Instrument trays, rectangular
340×510×50 mm
AGEN1252
:
Agencinox
Instrument trays, rectangular
AGEN1252
:
Agencinox
Udvid billedet
Instrument trays, rectangular
340×510×50 mm
Tilføj til kurv
1.280,00 kr
Måleenhed
1 * 1 Items
-
+
Tilføj til kurv
Stainless steel, 18/10.
Smooth surfaces for fast, easy cleaning
Good chemical resistance
Excellent thermal resistance
Seq:
02
W×D×H:
340×510×50 mm