Adapters, receiver, plain bent, Quickfit®
29/32 NS, 29/32 NS
Katalog # 271-0017
Leverandør: SciLabware
Gå til:
Product Details & Documents
Receiver, delivery, PYREX® borosilicate glass
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Plain bend without vent.
Specifikationer
- Cone size:29/32 NS
- Socket size:29/32 NS
- Seq:05