Order Entry
Denmark
Orders LinkContactUsLinkComponent
Culture shake flasks, WHEATON®
Culture shake flasks, WHEATON®
50 ml
Katalog # WHEA353255
Leverandør:   DWK Life Sciences
Culture shake flasks, WHEATON®
Katalog # WHEA353255
Leverandør:  DWK Life Sciences
Culture shake flasks, WHEATON®
Culture shake flasks, WHEATON®
50 ml
Price
1.100,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These culture shake flasks are made of 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
  • Baffles improve aeration and mixing for microbial and bacterial cultures
  • Break resistant lip for use with stopper, cotton plug or film cover
  • Pinched neck design to reduce splashing on flasks with four baffles
Designed for use with orbital or reciprocating shakers.
Specifikationer
  • Capacity:
    50 ml
  • Seq:
    01
Anbefalinger