Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

BOD bottles, WHEATON®

BOD bottles, borosilicate glass, 43×115 mm (Ø×H), un-numbered, 60 ml
Catalog # WHEA227494-00
Supplier:   DWK Life Sciences
BOD bottles, WHEATON®
Catalog # WHEA227494-00
Supplier:  DWK Life Sciences
BOD bottles, WHEATON®
BOD bottles, WHEATON®

BOD bottles, borosilicate glass, 43×115 mm (Ø×H), un-numbered, 60 ml

Price
16.300,00 kr
Log in for availability
1 * 36 Items
Product Details & Documents

Made of Type I borosilicate glass.

  • Available with or without stopper
  • With white labeling area
Specifications
  • Kapacitet:
    60 ml
  • Beskrivelse:
    BOD bottles, borosilicate glass, 43×115 mm (Ø×H), un-numbered