Order Entry
Denmark
Orders LinkContactUsLinkComponent
Funnels with conical joint
Funnels with conical joint
19/26 NS, 55 mm
Catalog # 221-0136
Supplier:   Lenz Laborglas
Funnels with conical joint
Catalog # 221-0136
Supplier:  Lenz Laborglas
Funnels with conical joint
Funnels with conical joint
19/26 NS, 55 mm
Price
200,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.
Documents
Specifications
  • Cone size:
    19/26 NS
  • Funnel O-Ø:
    55 mm