Order Entry
Denmark
Orders LinkContactUsLinkComponent
Samplers, Liquid-Sampler

Samplers, Liquid-Sampler

Liquid-Sampler, open with thumb, 32×540 mm, 100 ml
Catalog # 331-0033
Supplier:   BURKLE
Samplers, Liquid-Sampler
Catalog # 331-0033
Supplier:  BURKLE
Samplers, Liquid-Sampler
Samplers, Liquid-Sampler

Liquid-Sampler, open with thumb, 32×540 mm, 100 ml

Price
3.030,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Material: V4A stainless steel (1.4404). Sampler for thin to viscous liquids.

  • Easy one handed operation
  • Discharge funnel for safe filling of sample bottles

Documents

Specifications
  • Capacity:
    100 ml
  • Description:
    Liquid-Sampler, open with thumb, 32×540 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.