Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tripod stands
1,2 kg, 105 mm
  241-1109
 :   BOCHEM
Tripod stands
  241-1109
 :  BOCHEM
Tripod stands
Tripod stands
1,2 kg, 105 mm
635,00 kr
1 * 1 Items
Cast iron, varnished.
  • With M10 threaded hole
  • No support rod
  • Slip-proof, rubber feet ensure firm footing
  • Arm length:
    105 mm
  • Weight:
    1,2 kg