Order Entry
Denmark
Orders LinkContactUsLinkComponent
Funnels with conical joint
Funnels with conical joint
29/32 NS, 80 mm
Katalog # 221-0138
Leverandør:   Lenz Laborglas
Funnels with conical joint
Katalog # 221-0138
Leverandør:  Lenz Laborglas
Funnels with conical joint
Funnels with conical joint
29/32 NS, 80 mm
Price
329,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.
Documents
Specifikationer
  • Cone size:
    29/32 NS
  • Funnel O-Ø:
    80 mm