Order Entry
Denmark
Orders LinkContactUsLinkComponent
Balance brushes

Balance brushes

Natural bristle, 9,5 mm, 27 mm
Catalog # 611-9504
Marketsource
Balance brushes
Catalog # 611-9504
Marketsource
Balance brushes
Balance brushes

Natural bristle, 9,5 mm, 27 mm

Price
62,21 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Brushes made from fine bear hair.

Specifications
  • Bristle material:
    Natural bristle
  • Brush head Ø:
    9,5 mm
  • Trim length:
    27 mm