Order Entry
Denmark
Orders LinkContactUsLinkComponent
Multipurpose container, round, with snap-on lid

Multipurpose container, round, with snap-on lid

500 ml, 95,00 mm, 127,00 mm
Catalog # 216-3741
Supplier:   hünersdorff GmbH
Multipurpose container, round, with snap-on lid
Catalog # 216-3741
Supplier:  hünersdorff GmbH
Multipurpose container, round, with snap-on lid
Multipurpose container, round, with snap-on lid

500 ml, 95,00 mm, 127,00 mm

Price
37,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

HDPE, translucent.

  • Resistant to acids and chemicals
  • With tightly closing lid
  • Deep freezable
  • Stackable

For storing and transporting solid and liquid materials, samples and general labware.

Documents

Specifications
  • Ext.Ø:
    95,00 mm
  • Capacity:
    500 ml
  • Height:
    127,00 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.