Order Entry
Denmark
Orders LinkContactUsLinkComponent
Racks for test tubes

Racks for test tubes

18 mm, 24 (2×12), 320×60×80 mm
Catalog # 212-6340
Racks for test tubes
Catalog # 212-6340
Racks for test tubes
Racks for test tubes

18 mm, 24 (2×12), 320×60×80 mm

Price
3.240,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Rack for test tubes, made from clear acrylic, constructed in two tiers.

  • Good thermal resistance
  • Rugged stable design
  • Easy to clean
Specifications
  • Hole Ø:
    18 mm
  • No. of Holes:
    24 (2×12)
  • W×D×H:
    320×60×80 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.