Order Entry
Denmark
Orders LinkContactUsLinkComponent
Wipes for delicate tasks, Multisoft

Wipes for delicate tasks, Multisoft

White, 340×320 mm, Pack of 25
Catalog # 115-2726
Wipes for delicate tasks, Multisoft
Catalog # 115-2726
Wipes for delicate tasks, Multisoft
Wipes for delicate tasks, Multisoft

White, 340×320 mm, Pack of 25

Price
26,10 kr
Log in for availability
1 * 25 Items
Product Details & Documents

Soft non woven cloths made of Airlaid material, ideal for sensitive applications.

  • Solvent resistant and highly absorbent
  • Tear resistant, voluminous and lint-free
  • Packed in convenient dispensing box for single sheet dispensing
Specifications
  • Packed:
    Pack of 25
  • Colour:
    White
  • L×W:
    340×320 mm