Order Entry
Denmark
Orders LinkContactUsLinkComponent
Immunoassay microplates
Immunoassay microplates
Enhanced binding, flat bottom wells, Clear
Catalog # 735-0546
   Low Manufacturing Impact
Immunoassay microplates
Catalog # 735-0546
   Low Manufacturing Impact
Immunoassay microplates
Immunoassay microplates
Enhanced binding, flat bottom wells, Clear
Price
1.770,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents
Designed specifically for immunoassay applications, this range of products have binding surfaces that include Immulon®, universal binding (UB) and enhanced binding (EB). These make good experimental design easier, allowing development of assays with stringent pH, buffer, pKa and resin requirements.
  • Available in 96-well or strip plate format for matching to the requirements of automated instruments
  • Microplates with breakable strips offer maximum efficiency for processing both large and small sample numbers, as the exact number of wells required can be used
  • Break-apart wells have a unique well locking system that keeps the wells and samples securely in place
  • Strips available in 1×8 and 1×12 break-apart or solid strip format, with paired 2×8 strips also available
Immulon® 1 B and UB microplates have medium binding surfaces with a binding capacity of 100 to 200 ng IgG/cm². Both have an affinity for hydrophobic complexes and proteins. Molecules for medium binding surfaces include polyclonal antibodies, proteins, immunoglobulins, lipids, lipoproteins and lipopolysaccharides.
Immulon® 2 HB, Immulon® 4 HBX and EB microplates have high binding, irradiated surfaces, with a binding capacity of 400 to 500 ng IgG/cm². Immulon® 4 HBX microplates have an extra surface with special resin for maximum protein uptake. EB microplates provide guaranteed coefficient of variation. Molecules for high binding surfaces include monoclonal and polyclonal antibodies, proteins, peptides, polysaccharides, glycoproteins, immunoglobulins, lipoproteins, glycoproteins and purified antigens.
Specifications
  • Colour:
    Clear
  • Description:
    Enhanced binding, flat bottom wells
  • Seq:
    08
  • Cat. No.:
    735-0546
  • Environmental certification:
    ISO 14001 (Certificate Registration No. EMS 623400)
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Vinyl lid for 1×12 strips
    Vinyl lid for 1×12 strips
    735-0192
    Log in for availability
    1 * 100 Items - 1.570,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Holder for 1×12 strips
    Holder for 1×12 strips
    735-0193
    Log in for availability
    1 * 10 Items - 499,00 kr