Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kapselfiltre, Polycap™, Whatman™

Kapselfiltre, Polycap™, Whatman™

Polycap™ TC 36, PES, with filling bell, non sterile, 0,2/0,2 µm, MNPT/Stepped barb for 6 - 10 mm tubing
Catalog # 515-2538
Kapselfiltre, Polycap™, Whatman™
Catalog # 515-2538
Kapselfiltre, Polycap™, Whatman™
Kapselfiltre, Polycap™, Whatman™

Polycap™ TC 36, PES, with filling bell, non sterile, 0,2/0,2 µm, MNPT/Stepped barb for 6 - 10 mm tubing

Price
1.060,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

These Polycap™ TC capsule filters feature dual layer polytehersulfone (PES) membranes and provide efficient filtration for aqueous solutions.

  • 100% integrity-tested during manufacturing
  • PP housing thermally fused to eliminate surfactants and mold releasing agents that could affect analysis
  • Available in sterile and non sterile versions with a filling bell option
  • PES membranes protein adsorption characteristics: HAS 0,4 µg/cm²; Insulin 2,0 µg/cm²; gammaglobulin 1,5 µg/cm²

PES membranes are inherently hydrophilic, have low extractables, are biosafe, and have excellent flow rates. These characteristics make Polycap™ TC PES filters appropriate for a variety of critical filtration applications, including cleaning/rinsing pharmaceutical solutions, tissue culture media, and viral suspensions.

Polycap™ AS (Aqueous Solution) - recommended for filtering aqueous solutions. It combines a glass microfibre (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.

Polycap™ TF - designed for use with organic solvents and chemically aggressive solutions.

Polycap™ HD (Heavy Duty) - offers high filtration efficiency and excellent filtrate purity due to its materials and methods of manufacture.

Polycap™ SPF (Serum Prefilter) - optimised for prefiltration applications and typically used upstream of a Polycap™ AS or a Polycap™ PES capsule.

Certifications: Manufactured according to ISO9001:2008 QMS standards

Specifications
  • Tilslutning:
    MNPT/Stepped barb for 6 - 10 mm tubing
  • Porestørrelse:
    0,2/0,2 µm
  • Model:
    Polycap™ TC 36, PES, with filling bell, non sterile
  • Seq:
    04
  • Filtreringsareal:
    440 cm²
  • Cat. No.:
    515-2538
  • Maks. tryk:
    4,1 bar
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.