Order Entry
Denmark
Orders LinkContactUsLinkComponent
Single channel pipettes, electronic or manual, variable volume
Single channel pipettes, electronic or manual, variable volume
Piston lift pipette, 1 µl steps, KHP/Var 1000, 100 - 1000 µl
Katalog # 662-0431
Leverandør:   WTW
Single channel pipettes, electronic or manual, variable volume
Katalog # 662-0431
Leverandør:  WTW
Single channel pipettes, electronic or manual, variable volume
Single channel pipettes, electronic or manual, variable volume
Piston lift pipette, 1 µl steps, KHP/Var 1000, 100 - 1000 µl
Price
5.180,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The manually adjustable pipettes with variable volumes offer high flexibility and ease of use for photometric water analysis in the field and the lab. The electronic single channel pipettes with automatic volume pickup and volume output at the push of a button offers quick test throughput and even simpler operation.
  • Manually adjustable and robust
  • Electronic single channel pipettes adjustable for high test throughput
Specifikationer
  • Volume:
    100 - 1000 µl
  • Description:
    Piston lift pipette, 1 µl steps, KHP/Var 1000
Anbefalinger