Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

IgM purification kit , Pierce™

MBP Agarose, mannan binding protein on 4% beaded agarose, 10 mL
Catalog # PIER22212
IgM purification kit , Pierce™
Catalog # PIER22212
CAS Number:  
IgM purification kit , Pierce™
IgM purification kit , Pierce™

MBP Agarose, mannan binding protein on 4% beaded agarose, 10 mL

CAS Number:  
Price
13.810,00 kr
Log in for availability
1 * 10 mL
Product Details & Documents

The Pierce™ IgM Purification Kit is an easy to use affinity matrix and buffer system to purify IgM from mouse ascites fluid. The method uses immobilised mannan binding protein (MBP), which binds to IgM in the kit-supplied buffers.

  • Immobilised MBP allows specific purification of mouse IgM from ascites fluid
  • Non denaturing, near-neutral buffer conditions ensure recovery of functional antibodies
  • Stable ligand and resins can be regenerated and reused at least ten times

 : Kit includes MBP Agarose column, 5 m, 1 column; IgM Binding buffer, 800 ml; IgM Elution buffer, 500 ml; MBP Column Preparation buffer, 50 ml; and Column extender.

Specifications

Specifications

  • Pk:
    10 mL
  • Description:
    MBP Agarose, mannan binding protein on 4% beaded agarose

Specifications