Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spider evaporators
Spider evaporators
Spider with evaporating flasks, SJ29/32, glass, 100 ml, For simultaneous distillation from 5×100 ml evaporating flasks with STJ 24/29
Catalog # 531-1320
Supplier:   BUCHI
Spider evaporators
Catalog # 531-1320
Supplier:  BUCHI
Spider evaporators
Spider evaporators
Spider with evaporating flasks, SJ29/32, glass, 100 ml, For simultaneous distillation from 5×100 ml evaporating flasks with STJ 24/29
Price
10.150,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Specifications
  • Description:
    Spider with evaporating flasks, SJ29/32, glass, 100 ml
  • Type:
    For simultaneous distillation from 5×100 ml evaporating flasks with STJ 24/29