Microplates, 384-well
Microplate, PP, flat bottom, deep shape, 384-well, with loose glass inserts, chemically inert, ~250 µl
Catalog # 612-9873
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Highly pure, chemically inert aluminium plate for individual glass inserts. Translucent plastic cover included.
Specifications
- Description:Microplate, PP, flat bottom, deep shape, 384-well, with loose glass inserts, chemically inert
- Well volume:~250 µl
Accessories
Product Information
- Catalog #AvailabilityPrice
- Glass inserts for 384 well plates