Order Entry
Denmark
Orders LinkContactUsLinkComponent
Apparatus - Antonacopoulos
Apparatus - Antonacopoulos
Complete apparatus, - NS, - NS
Catalog # LENZ05434000
Supplier:   Lenz Laborglas
Apparatus - Antonacopoulos
Catalog # LENZ05434000
Supplier:  Lenz Laborglas
Apparatus - Antonacopoulos
Complete apparatus, - NS, - NS
Price
6.100,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Documents
Specifications
  • Cone size:
    - NS
  • Socket size:
    - NS
  • Description:
    Complete apparatus
  • Seq:
    01