Order Entry
Denmark
Orders LinkContactUsLinkComponent
O-rings for flat flanges, DURAN®

O-rings for flat flanges, DURAN®

Flat flanges DN 150, Red, 157 mm
Catalog # 201-0164
Supplier:   DWK Life Sciences
O-rings for flat flanges, DURAN®
Catalog # 201-0164
Supplier:  DWK Life Sciences
O-rings for flat flanges, DURAN®
O-rings for flat flanges, DURAN®

Flat flanges DN 150, Red, 157 mm

Price
689,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Silicone core sheathed with FEP, red.

  • Guarantee excellent joint seal
  • Temperature resistant from −200 to +200 °C
  • Excellent chemical resistance

O-rings for flat flanges with groove.

Specifications
  • For:
    Flat flanges DN 150
  • Ø int.:
    157 mm
  • Colour:
    Red
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.