Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
4 mm, 18,8 NS
Catalog # SCOT286440405
Supplier:   DWK Life Sciences
Stopcocks, single way, DURAN®
Catalog # SCOT286440405
Supplier:  DWK Life Sciences
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
4 mm, 18,8 NS
Price
4.420,00 kr
Log in for availability
1 * 25 Items
Product Details & Documents
DURAN®, borosilicate glass 3,3, clear, polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
  • For glass manipulators
Ungrounded, without key.
Specifications
  • Hole Ø:
    4 mm
  • Spigot:
    18,8 NS