Stopcocks, single-way, DURAN®
6 mm, 21,5 NS
SCOT286414601
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
- Hole Ø:6 mm
- Seq:30
- Spigot:21,5 NS
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
Straight bore.