Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stopcocks, single-way, DURAN®
Stopcocks, single-way, DURAN®
2,5 mm, 14,5 NS
Catalog # SCOT286412009
Supplier:   DWK Life Sciences
Stopcocks, single-way, DURAN®
Catalog # SCOT286412009
Supplier:  DWK Life Sciences
Stopcocks, single-way, DURAN®
Stopcocks, single-way, DURAN®
2,5 mm, 14,5 NS
Price
10.090,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.
Specifications
  • Hole Ø:
    2,5 mm
  • Seq:
    16
  • Spigot:
    14,5 NS