Cleaning solution DZ 8050 for gentle removal of lime residues 1 * 1 L
Catalog # SCOI285530276
Supplier: SI Analytics
Cleaning solution DZ 8050 for gentle removal of lime residues 1 * 1 L
Catalog # SCOI285530276
Supplier: SI Analytics
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals