Order Entry
Denmark
ContactUsLinkComponent
Connecting cable VZ 7114 (insert frame VZ 7003 insert frame VZ 7003). This connection cable is required for arrangements with mo re than 4 measuring positions. 1 * 1 items
Catalog # SCOI285421195
Supplier:  SI Analytics
Connecting cable VZ 7114 (insert frame VZ 7003 insert frame VZ 7003). This connection cable is required for arrangements with mo re than 4 measuring positions. 1 * 1 items
Catalog # SCOI285421195
Supplier:  SI Analytics
Frequently Bought Together