Order Entry
Denmark
ContactUsLinkComponent
pH combination electrode L 7781 HD with screw plug head. Low maintenance, glass shaft, ceramic diaphragm, gel electrolyte, Silamid®- reference system, sphere membrane, L-glass, length 120 mm, 12 mm Ø, -5...+60 °C, 2...12 pH 1 * 1 items
Catalog # 662-2276
Supplier:  SI Analytics
pH combination electrode L 7781 HD with screw plug head. Low maintenance, glass shaft, ceramic diaphragm, gel electrolyte, Silamid®- reference system, sphere membrane, L-glass, length 120 mm, 12 mm Ø, -5...+60 °C, 2...12 pH 1 * 1 items
Catalog # 662-2276
Supplier:  SI Analytics
Frequently Bought Together