Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Filter discs, DURAN®

3, 90 mm, 6 mm
  SCOT251590302
Filter discs, DURAN®
  SCOT251590302
Filter discs, DURAN®
Filter discs, DURAN®

3, 90 mm, 6 mm

 
324,00 kr
   
1 * 1 Items
 

Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.

  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design

Made from clear DURAN® borosilicate glass 3,3.

 
  • Ø:
    90 mm
  • Porosity:
    3
  • Seq:
    66
  • Thickness:
    6 mm