Order Entry
Denmark
Orders LinkContactUsLinkComponent
Funnel stands

Funnel stands

Plates with aluminum support rod (Ø×L: 12,7×595 mm), 250×140 mm, 2 funnels
Catalog # 519-2711
Supplier:   Brand
Funnel stands
Catalog # 519-2711
Supplier:  Brand
Funnel stands
Funnel stands

Plates with aluminum support rod (Ø×L: 12,7×595 mm), 250×140 mm, 2 funnels

Price
1.770,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

PP plates for holding two or four funnels with top outer-Ø of 50 to 120 mm.

  • With aluminum support rod (Ø×L: 12,7×595 mm)
  • Adjustable funnel height

Documents

Specifications
  • For:
    2 funnels
  • Description:
    Plates with aluminum support rod (Ø×L: 12,7×595 mm)
  • L×W:
    250×140 mm