Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
25 L
Supplier: Avantor
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 7053.9025
Supplier: Avantor
CAS Number: 75-09-2
Jump to:
Product Details & Documents
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Boiling Pt:39.8 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:1.322 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-838-9
- UN:1593
- ADR:6.1,III
Documents
Certificates
Search by Lot Numberspec_BAKR7053.1000.pdf spec_BAKR7053.2500.pdf spec_BAKR7053.9025.pdf
Product specification
Specifications
Specification Test Results
- Assay (by GC) (corrected for water):min. 99,5 %
- Appearance:clear
- Color (APHA):max. 10
- Free Halogens:passes test
- Residue after Evaporation:max. 0,002 %
- Titrable Acid (meq/g):max. 0,0002
- Water (H2O):max. 0,02 %
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Specifications
- Pk:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



