Order Entry
Denmark
Orders LinkContactUsLinkComponent
Weighing scoops

Weighing scoops

90 mm, 32 mm
Catalog # BOCH12811
Supplier:   BOCHEM
Weighing scoops
Catalog # BOCH12811
Supplier:  BOCHEM
Weighing scoops
Weighing scoops

90 mm, 32 mm

Price
60,50 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Aluminium.

Specifications
  • Width:
    32 mm
  • Length:
    90 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.