Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Baumé hydrometers

0 - 20 °Bé, 0,1, 330 mm
  AMARH842876
 :   Amarell
Baumé hydrometers
  AMARH842876
 :  Amarell
Baumé hydrometers
Baumé hydrometers

0 - 20 °Bé, 0,1, 330 mm

 
275,00 kr
   
1 * 1 Items
 

General purpose Baumé hydrometers with or without thermometer.

  • Reference temperature of 15 °C
  • Paper scale with visible print
  • Shot weighted body

Colourless round capillary with red or blue filling.

 
  • Length:
    330 mm
  • Seq:
    06
  • Division:
    0,1
  • Range:
    0 - 20 °Bé