Special red felt pen,neutral labels for individual labeling made of white pvc film. with very good adhesion capacity and rounded cor ners. insensitive to acids, alkalis and alcohol red felt pen 1 * 1 items
811-1251
: SEMADENI
Frequently Bought Together