Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pycnometers, unadjusted
Pycnometers, unadjusted
50 ml
Catalog # 613-2177
Supplier:   witeg Labortechnik
Pycnometers, unadjusted
Catalog # 613-2177
Supplier:  witeg Labortechnik
Pycnometers, unadjusted
Pycnometers, unadjusted
50 ml
Price
277,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in capillary stopper
  • Nominal capacity printed on the bottom
  • Very good chemical and temperature resistance
Certifications: DIN 12797
Specifications
  • Volume (nom.):
    50 ml
Anbefalinger