Order Entry
Denmark
Orders LinkContactUsLinkComponent
Column end plugs
Column end plugs
Column end plug, black
Catalog # 554-1146
Supplier:   VICI JOUR RESEARCH
Column end plugs
Catalog # 554-1146
Supplier:  VICI JOUR RESEARCH
Column end plugs
Column end plugs
Column end plug, black
Price
155,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
These nylon column end plugs fit all columns with 10-32 threads. The long design and threading down to the ferrule ensure safe closing of valuable columns.
  • Available with a 5/16" wrench flat
  • Easy removal
Specifications
  • Description:
    Column end plug, black
  • Seq:
    02