Hydroxylammonium sulphate ≥99%
Glass bottle for solids, 2 kg
Supplier: Thermo Fisher Scientific
Hydroxylammonium sulphate ≥99%
Catalog # 88944.A3
Supplier: Thermo Fisher Scientific
CAS Number: 10039-54-0
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 10039-54-0 Molecular Formula: H3NO.0.5H2SO4 Formula Weight: 164.14 Physical Form: Crystalline Density: 1.88 Appearance: White Odour: Odourless pH: 3.6 Melting Point: ca 170° dec. Solubility: Soluble in water Merck Reference: 14,4828 UN No.: 2865 MDL No.: MFCD00044869
Synonyms: Bis(hydroxylammonium) sulphate, Hydroxylamine sulphate, Hydroxylamine sulphate (2:1)
- CAS Number:10039-54-0
- Formula:H₈N₂O₆S
- MDL Number:MFCD00044869
- Molecular Weight:164.14 g/mol
- Merck Index:14,4828
- Melting Pt:∼ 170 °C (dec.)
- Density:1.88
- Storage Temperature:Ambient
- EINECS:233-118-8
- UN:2865
- ADR:8,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



