Stirrer magnetic micro 7x2mm,micro-magnetic stirrers - ptfewith permanent magnetic core and thick, white ptfe coating. chemical and temperature resistant up to +275 ° c. 7x2 mm 1 * 1 items
Catalog # 442-9333
Supplier: SEMADENI
Frequently Bought Together