Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Retort Rods
Stand rods
Stand rods
Print
Stand rods
12 mm, 1250 mm
Catalog # 241-7171
Supplier:
USBECK Laborgeräte
Stand rods
Catalog # 241-7171
Supplier:
USBECK Laborgeräte
Expand image
Return to Top
Stand rods
12 mm, 1250 mm
Specifications
Add to cart
Jump to:
Specifications
Price
295,00 kr
Log in
for availability
Unit of Measure
1 * 1 Items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Stainless steel.
Robust, very stable
Excellent chemical resistance
Specifications
Ø:
12 mm
Length:
1250 mm
Seq:
15