Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tripod stands

Tripod stands

2,0 kg, 145 mm
Catalog # 241-7123
Supplier:   BOCHEM
Tripod stands
Catalog # 241-7123
Supplier:  BOCHEM
Tripod stands
Tripod stands

2,0 kg, 145 mm

Price
695,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Cast iron, varnished.

  • With M10 threaded hole
  • No support rod
  • Slip-proof, rubber feet ensure firm footing
Specifications
  • Arm length:
    145 mm
  • Weight:
    2,0 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.