Order Entry
Denmark
Orders LinkContactUsLinkComponent
Double spatulas, micro

Double spatulas, micro

210 mm, 60 mm, 9 mm
Catalog # 231-2189
Supplier:   BOCHEM
Double spatulas, micro
Catalog # 231-2189
Supplier:  BOCHEM
Double spatulas, micro
Double spatulas, micro

210 mm, 60 mm, 9 mm

Price
30,40 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stainless steel 18/10, anti-magnetic. With round handle, one flat and one scoop shaped, rounded spatula end.

  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Specifications
  • Length:
    210 mm
  • Blade width:
    9 mm
  • Blade length:
    60 mm